Invitrogen
Manufacturer Code:PA521049
Catalog # PIPA521049
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide near the carboxy terminus of human MFSD2A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: major facilitator superfamily domain containing 2; Major facilitator superfamily domain-containing protein 2A; mfsd2a zgc:101615 major facilitator superfamily domain containing 2a major facilitator superfamily domain-containing protein 2A-A; NLS1; sodium-dependent LPC symporter 1; Sodium-dependent lysophosphatidylcholine symporter 1
Gene Aliases: 1700018O18Rik; HMFN0656; MCPH15; MFSD2; MFSD2A; NLS1; PP9177; RGD1310174; UNQ300/PRO341
UniProt ID: (Human) Q8NA29, (Mouse) Q9DA75
Entrez Gene ID: (Human) 84879, (Rat) 298504, (Mouse) 76574
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.