Novus Biologicals
Manufacturer Code:NBP281929
Catalog # NB017240
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MFSD2A antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human MFSD2A. The immunogen is located within the last 50 amino acids of MFSD2A. Amino Acid Squence: PIDEERRRQNKKALQALR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ14490 FLJ35904 major facilitator superfamily domain containing 2 major facilitator superfamily domain containing 2A major facilitator superfamily domain-containing protein 2A MFSD2; Major facilitator superfamily domain-containing protein 2A; NLS1; sodium-dependent LPC symporter 1; Sodium-dependent lysophosphatidylcholine symporter 1
Gene Aliases: HMFN0656; MCPH15; MFSD2; MFSD2A; NLS1; PP9177; UNQ300/PRO341
UniProt ID: (Human) Q8NA29
Entrez Gene ID: (Human) 84879
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.