Proteintech
Manufacturer Code:254981AP150UL
Catalog # 50-173-4932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MFSD2 Fusion Protein Ag22232 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FP1147 HMFN0656 MFSD2 MFSD2A PP10484 PP9177 RP3 342P20.1 UNQ300/PRO341; Major facilitator superfamily domain-containing protein 2A; NLS1; sodium-dependent LPC symporter 1; Sodium-dependent lysophosphatidylcholine symporter 1
Gene Aliases: HMFN0656; MCPH15; MFSD2; MFSD2A; NLS1; PP9177; UNQ300/PRO341
UniProt ID: (Human) Q8NA29
Entrez Gene ID: (Human) 84879
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.