Invitrogen
Manufacturer Code:MA527647
Catalog # PIMA527647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S153-5 |
Immunogen | Fusion protein amino acids 370-600 (cytoplasmic N-terminus) of mouse Mitofusin-2. Rat: 97&percnt identity (226/231 amino acids identical). Human: 92&percnt identity (214/231 amino acids identical) &bsim55&percnt identity with Mitofusin-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CMT2A CMT2A2 CPRP 1 CPRP1 Fzo HSG hyperplasia suppressor gene Hypertension related protein 1 KIAA0214 MARF MFN 2 Mfn2 Mitochondrial assembly regulatory factor Mitofusin-2 Mitofusin2 Transmembrane GTPase MFN2; HSG protein; hyperplasia suppressor; hypertension related protein 1; hypertension-related protein; Hypertension-related protein 1; Mitochondrial assembly regulatory factor; mitochondrial transmembrane GTPase FZO1A; Mitofusin-2; Transmembrane GTPase MFN2
Gene Aliases: CMT2A; CMT2A2; CPRP1; D630023P19Rik; Fzo; HMSN6A; HSG; KIAA0214; MARF; MFN2
UniProt ID: (Human) O95140, (Mouse) Q80U63
Entrez Gene ID: (Human) 9927, (Mouse) 170731, (Rat) 64476
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.