Abnova
Manufacturer Code:PAB31349
Catalog # 89-998-305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human MFI2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5; CD228; FLJ38863; MAP97; Melanoma-associated antigen p97; melanoma-associated antigen p97, isoform 2; Melanotransferrin; membrane-bound transferrin-like protein; MGC4856; MTF1
Gene Aliases: CD228; MAP97; MELTF; MFI2; MTf; MTF1
UniProt ID: (Human) P08582
Entrez Gene ID: (Human) 4241
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.