Proteintech
Manufacturer Code:104281AP150UL
Catalog # 50-172-6301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MFI2 Fusion Protein Ag0700 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5; CD228; CD228 MAP97 Melanotransferrin MFI2 MTF1; Melanoma-associated antigen p97; melanoma-associated antigen p97, isoform 2; Melanotransferrin; membrane-bound transferrin-like protein; Membrane-bound transferrin-like protein p97; MTf
Gene Aliases: CD228; MAP97; MELTF; MFI2; MTf; MTF1
UniProt ID: (Human) P08582, (Mouse) Q9R0R1
Entrez Gene ID: (Human) 4241, (Mouse) 30060, (Rat) 288038
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.