R&D Systems
Manufacturer Code:IC27671T100UG
Catalog # IC27671T100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 278918 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human MFG-E8 Leu24-Cys387 Accession # Q08431 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: BA46 Breast epithelial antigen BA46 EDIL1 hP47 HsT19888 lactadherin Lactahedrin Medin MFG1 MFGE8 MFG-E8 MFGM Milk fat globule-EGF factor 8 milk fat globule-EGF factor 8 protein O-acetyl disialoganglioside synthase OAcGD3S SED1 SPAG10 sperm associated antigen 10 sperm surface protein hP47; Breast epithelial antigen BA46; HMFG; Lactadherin; Lactadherin short form; Medin; MFG-E8; MFGM; Milk fat globule-EGF factor 8; O-acetyl disialoganglioside synthase; SED1; sperm associated antigen 10; sperm surface protein hP47
Gene Aliases: BA46; EDIL1; HMFG; hP47; HsT19888; MFG-E8; MFGE8; MFGM; OAcGD3S; SED1; SPAG10
UniProt ID: (Human) Q08431
Entrez Gene ID: (Human) 4240
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.