Proteintech
Manufacturer Code:170901AP150UL
Catalog # 50-561-082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MFF Fusion Protein Ag9990 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C2orf33 MFF mitochondrial fission factor; Mitochondrial fission factor
Gene Aliases: 5230400G24Rik; AD030; AD033; AI314724; C2orf33; GL004; MFF; RGD1310230
UniProt ID: (Human) Q9GZY8, (Rat) Q4KM98, (Mouse) Q6PCP5
Entrez Gene ID: (Human) 56947, (Rat) 301563, (Mouse) 75734
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.