Invitrogen
Manufacturer Code:PA5100206
Catalog # PIPA5100206
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MFAP5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAT9 MAGP2 MAGP-2 MFAP5 MFAP-5 Microfibril-associated glycoprotein 2 microfibril-associated glycoprotein-2 microfibrillar associated protein 5 microfibrillar-associated protein 5 MP25 THE1A-MFAP5; MAGP-2; MFAP-5; Microfibril-associated glycoprotein 2; microfibril-associated glycoprotein-2; Microfibrillar-associated protein 5; MP25; THE1A-MFAP5
Gene Aliases: AAT9; MAGP-2; MAGP2; MFAP-5; MFAP5; MP25
UniProt ID: (Human) Q13361, (Mouse) Q9QZJ6
Entrez Gene ID: (Human) 8076, (Rat) 362429, (Mouse) 50530
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.