Proteintech
Manufacturer Code:176611AP150UL
Catalog # 50-173-1709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MFAP4 Fusion Protein Ag11920 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MFAP4; Microfibril-associated glycoprotein 4; microfibrillar-associated protein 4
Gene Aliases: 1110007F23Rik; Magp-36; MFAP4
UniProt ID: (Human) P55083, (Mouse) Q9D1H9
Entrez Gene ID: (Human) 4239, (Mouse) 76293, (Rat) 287382
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.