Proteintech
Manufacturer Code:167911AP150UL
Catalog # 50-173-1182
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | METTL5 Fusion Protein Ag10232 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPC133 methyltransferase like 5 METTL5; Methyltransferase-like protein 5; rRNA N6-adenosine-methyltransferase METTL5
Gene Aliases: 2810410A08Rik; DC3; HSPC133; METTL5; RGD1566062
UniProt ID: (Human) Q9NRN9, (Mouse) Q8K1A0
Entrez Gene ID: (Human) 29081, (Mouse) 75422, (Rat) 502632
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.