Invitrogen
Manufacturer Code:PA569560
Catalog # PIPA569560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human METTL4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HsT661 methyltransferase-like protein 4; Methyltransferase-like protein 4; N(6)-adenine-specific DNA methyltransferase METTL4; N(6)-adenine-specific methyltransferase METTL4; snRNA (2'-O-methyladenosine-N(6)-)-methyltransferase METTL4
Gene Aliases: 2410198H06Rik; A730091E08Rik; AV296509; HsT661; METTL4; RGD1306451
UniProt ID: (Human) Q8N3J2, (Mouse) Q3U034
Entrez Gene ID: (Human) 64863, (Sheep) 100913156, (Rabbit) 100343651, (Bovine) 521222, (Dog) 490534, (Horse) 100061890, (Mouse) 76781, (Rat) 316731, (Guinea pig) 100730430, (Zebrafish) 560686
Molecular Function:
DNA binding protein
DNA methyltransferase
RNA binding protein
RNA methyltransferase
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.