Novus Biologicals
Manufacturer Code:NBP30449020UL
Catalog # NB094330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human METTL4 (NP_073751.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1.- FLJ23017 HsT661 methyltransferase like 4 methyltransferase-like protein 4 MGC117235; Methyltransferase-like protein 4; N(6)-adenine-specific DNA methyltransferase METTL4; N(6)-adenine-specific methyltransferase METTL4; snRNA (2'-O-methyladenosine-N(6)-)-methyltransferase METTL4
Gene Aliases: HsT661; METTL4
UniProt ID: (Human) Q8N3J2
Entrez Gene ID: (Human) 64863
Molecular Function: DNA binding protein DNA methyltransferase RNA binding protein RNA methyltransferase methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.