Abnova
Manufacturer Code:H00064863B01P
Catalog # 89-107-296
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | METTL4 (NP_073751.21 a.a. &bsim 472 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human METTL4 protein.
For Research Use Only
Protein Aliases: FLJ23017; HsT661; Methyltransferase-like protein 4; MGC117235; N(6)-adenine-specific DNA methyltransferase METTL4; N(6)-adenine-specific methyltransferase METTL4; snRNA (2'-O-methyladenosine-N(6)-)-methyltransferase METTL4
Gene Aliases: HsT661; METTL4
UniProt ID: (Human) Q8N3J2
Entrez Gene ID: (Human) 64863
Molecular Function:
DNA binding protein
DNA methyltransferase
RNA binding protein
RNA methyltransferase
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.