Novus Biologicals
Manufacturer Code:NBP187995
Catalog # NBP187995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:PYWAIYWPGGQALSRYLLDNPDVVRGKSVLDLGSGCGATAIAAKMSGASRILANDIDPIAGMAITLNCELNRLNP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C12orf72 chromosome 12 open reading frame 72 DKFZp451L235 EC 2.1.1.- hypothetical protein LOC254013 methyltransferase like 20 MGC50559; Electron transfer flavoprotein beta subunit lysine methyltransferase; ETFB lysine methyltransferase; ETFB-KMT; methyltransferase like 20; methyltransferase-like protein 20; Protein N-lysine methyltransferase METTL20
Gene Aliases: C12orf72; ETFB-KMT; ETFBKMT; METTL20
UniProt ID: (Human) Q8IXQ9
Entrez Gene ID: (Human) 254013
Molecular Function: DNA binding protein DNA methyltransferase RNA binding protein RNA methyltransferase methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.