Novus Biologicals
Manufacturer Code:NBP304387100UL
Catalog # NB093755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 105-262 of human METTL20 (NP_001129335.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C12orf72 chromosome 12 open reading frame 72 DKFZp451L235 EC 2.1.1.- hypothetical protein LOC254013 methyltransferase like 20 MGC50559; Electron transfer flavoprotein beta subunit lysine methyltransferase; ETFB lysine methyltransferase; ETFB-KMT; methyltransferase like 20; methyltransferase-like protein 20; Protein N-lysine methyltransferase METTL20
Gene Aliases: C12orf72; ETFB-KMT; ETFBKMT; METTL20
UniProt ID: (Human) Q8IXQ9
Entrez Gene ID: (Human) 254013
Molecular Function:
DNA binding protein
DNA methyltransferase
RNA binding protein
RNA methyltransferase
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.