Invitrogen
Manufacturer Code:PA543606
Catalog # PIPA543606
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human KIAA1627 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hMETTL14; Methyltransferase-like protein 14; N6-adenosine-methyltransferase non-catalytic subunit; N6-adenosine-methyltransferase subunit METTL14; snoRNA MBII-84
Gene Aliases: G430022H21Rik; KIAA1627; METTL14; mKIAA1627; RGD1304822; zgc:77296
UniProt ID: (Human) Q9HCE5, (Bovine) A4IFD8, (Mouse) Q3UIK4, (Zebrafish) Q6NZ22
Entrez Gene ID: (Human) 57721, (Bovine) 531382, (Dog) 487920, (Horse) 100072440, (Rabbit) 100340084, (Pig) 100525761, (Mouse) 210529, (Rat) 295428, (Zebrafish) 404603
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.