Invitrogen
Manufacturer Code:PA5117138
Catalog # PIPA5117138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human METTL14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G430022H21Rik hMETTL14 Kiaa1627 methyltransferase like 14 methyltransferase-like protein 14 Mettl14 mKIAA1627 N6-adenosine-methyltransferase non-catalytic subunit N6-adenosine-methyltransferase subunit METTL14 RGD1304822 snoRNA MBII-84; hMETTL14; Methyltransferase-like protein 14; N6-adenosine-methyltransferase non-catalytic subunit; N6-adenosine-methyltransferase subunit METTL14; snoRNA MBII-84
Gene Aliases: G430022H21Rik; KIAA1627; METTL14; mKIAA1627; RGD1304822
UniProt ID: (Human) Q9HCE5, (Mouse) Q3UIK4
Entrez Gene ID: (Human) 57721, (Mouse) 210529, (Rat) 295428
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.