Novus Biologicals
Manufacturer Code:NBP297952C
Catalog # NB049862
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human METTL14 fragment. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1 EC 2.1.1.- KIAA1627methyltransferase-like protein 14 methyltransferase like 14; hMETTL14; Methyltransferase-like protein 14; N6-adenosine-methyltransferase non-catalytic subunit
Gene Aliases: KIAA1627; METTL14
UniProt ID: (Human) Q9HCE5
Entrez Gene ID: (Human) 57721
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.