Proteintech
Manufacturer Code:261581AP150UL
Catalog # 50-173-5239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | METTL14 Fusion Protein Ag14325 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hMETTL14; KIAA1627 methyltransferase like 14 METTL14; Methyltransferase-like protein 14; N6-adenosine-methyltransferase non-catalytic subunit
Gene Aliases: KIAA1627; METTL14
UniProt ID: (Human) Q9HCE5
Entrez Gene ID: (Human) 57721
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.