Proteintech
Manufacturer Code:149941AP150UL
Catalog # 50-172-9895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | METTL1 Fusion Protein Ag6980 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C12orf1 methyltransferase like 1 METTL1 TRM8 tRNA(m7G46) methyltransferase YDL201w; D1075-like gene product; methyltransferase like 1; methyltransferase-like 1; Methyltransferase-like protein 1; METTL1; miRNA (guanine-N(7)-)-methyltransferase; mRNA (guanine-N(7)-)-methyltransferase; tRNA (guanine(46)-N(7))-methyltransferase; tRNA (guanine-N(7)-)-methyltransferase; tRNA(m7G46)-methyltransferase
Gene Aliases: 2810012D02Rik; C12orf1; METTL1; TRM8; TRMT8; YDL201w
UniProt ID: (Human) Q9UBP6, (Mouse) Q9Z120
Entrez Gene ID: (Human) 4234, (Rat) 679091, (Mouse) 17299
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.