OriGene
Manufacturer Code:TA504728
Catalog # 50-167-5351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A2 |
Immunogen | Full length human recombit protein of human METT10D produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: methyltransferase 10 domain containing; Methyltransferase 10 domain-containing protein; Methyltransferase-like protein 16; METT10D; N6-adenosine-methyltransferase METTL16; putative methyltransferase METT10D; RNA N6-adenosine-methyltransferase METTL16; U6 small nuclear RNA (adenine-(43)-N(6))-methyltransferase
Gene Aliases: METT10D; METTL16
UniProt ID: (Human) Q86W50
Entrez Gene ID: (Human) 79066
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.