OriGene
Manufacturer Code:CF800289
Catalog # 50-166-5001
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F6 |
Immunogen | Human recombit protein fragment corresponding to amino acids 2-212 of human METAP2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: eIF-2-associated p67 homolog; Initiation factor 2-associated 67 kDa glycoprotein; MAP 2; MAP2 MNPEP p67eIF2; Methionine aminopeptidase 2; p67; Peptidase M; peptidase M 2; testicular tissue protein Li 17
Gene Aliases: MAP2; METAP2; MNPEP; P67EIF2
UniProt ID: (Human) P50579
Entrez Gene ID: (Human) 10988
Molecular Function:
hydrolase
metalloprotease
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.