Invitrogen
Manufacturer Code:MA538071
Catalog # PIMA538071
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2022 |
Immunogen | A synthesized peptide derived from human METAP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 4930584B20Rik A930035J23Rik AI047573 AL024412 Amp2 AU014659 eIF-2-associated p67 eIF-2-associated p67 homolog initiation factor 2 associated 67 kDa protein initiation factor 2-associated 67 kDa glycoprotein initiation factor 2-associated protein (p67) MAP 2 MAP2 metAP 2 METAP2 Methionine aminopeptidase 2 methionyl aminopeptidase 2 Mnpep p67 p67eIF2 Peptidase M peptidase M 2 similar to methionyl aminopeptidase 2 testicular tissue protein Li 17; eIF-2-associated p67; eIF-2-associated p67 homolog; initiation factor 2 associated 67 kDa protein; Initiation factor 2-associated 67 kDa glycoprotein; initiation factor 2-associated protein (p67); MAP 2; metAP 2; Methionine aminopeptidase 2; p67; p67eIF2; Peptidase M; peptidase M 2; testicular tissue protein Li 17
Gene Aliases: 4930584B20Rik; A930035J23Rik; AI047573; AL024412; Amp2; AU014659; MAP2; METAP2; MNPEP; p67; P67EIF2
UniProt ID: (Human) P50579, (Mouse) O08663, (Rat) P38062
Entrez Gene ID: (Human) 10988, (Mouse) 56307, (Rat) 64370
Molecular Function:
hydrolase
metalloprotease
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.