Invitrogen
Manufacturer Code:MA530669
Catalog # PIMA530669
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G7E1H7 |
Immunogen | Recombit Human MAP1D protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CDS of metAP-3 within PCR fragment; MAP 1D; MAP1D Metap1l CDS of metAP-3 within PCR fragment methionine aminopeptidase 1D mitochondrial methionyl aminopeptidase type 1D mitochondrial; metAP 1D; Methionine aminopeptidase 1D, mitochondrial; Methionyl aminopeptidase type 1D, mitochondrial; Peptidase M 1D
Gene Aliases: MAP 1D; MAP1D; MetAP 1D; METAP1D; Metap1l
UniProt ID: (Human) Q6UB28
Entrez Gene ID: (Human) 254042
Molecular Function:
hydrolase
metalloprotease
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.