Invitrogen
Manufacturer Code:PA524339
Catalog # PIPA524339
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 306-334 amino acids from the N-terminal region of human MAP1D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDS of metAP-3 within PCR fragment; MAP 1D; MAP1D Metap1l CDS of metAP-3 within PCR fragment methionine aminopeptidase 1D mitochondrial methionyl aminopeptidase type 1D mitochondrial; metAP 1D; Methionine aminopeptidase 1D, mitochondrial; methionine aminopeptidase-like 1; Methionyl aminopeptidase type 1D, mitochondrial; Peptidase M 1D
Gene Aliases: 2310066F24Rik; 3110033D18Rik; AV117938; MAP 1D; MAP1D; MetAP 1D; METAP1D; Metap1l; Metapl1; Mnpepl
UniProt ID: (Human) Q6UB28, (Mouse) Q9CPW9
Entrez Gene ID: (Human) 254042, (Mouse) 66559
Molecular Function:
hydrolase
metalloprotease
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.