Invitrogen
Manufacturer Code:PA558203
Catalog # PIPA558203
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to Human METAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Store at &minus20°C long term. Avoid freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAP 1; MAP1 MetAP1 DKFZp781C0419 EC 3.4.11.18 KIAA0094MAP 1 MAP1A MetAP 1 MetAP1A methionine aminopeptidase 1 methionyl aminopeptidase 1 Peptidase M 1; metAP 1; Methionine aminopeptidase 1; Peptidase M 1
Gene Aliases: KIAA0094; MAP1A; METAP1; MetAP1A
UniProt ID: (Human) P53582
Entrez Gene ID: (Human) 23173
Molecular Function:
hydrolase
metalloprotease
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.