Invitrogen
Manufacturer Code:PA544285
Catalog # PIPA544285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human METAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAP 1; metAP 1; Methionine aminopeptidase 1; Peptidase M 1; Putative methionine aminopeptidase 1 (MetAP 1) (Peptidase M 1)
Gene Aliases: 1700029C17Rik; AW047992; fc84e12; im:7047238; KIAA0094; MAP1A; METAP1; MetAP1A; mKIAA0094; wu:fc84e12; zgc:110093
UniProt ID: (Human) P53582, (Bovine) A6QLA4, (Mouse) Q8BP48
Entrez Gene ID: (Dog) 487871, (Rabbit) 100351430, (Human) 23173, (Horse) 100066296, (Bovine) 516540, (Mouse) 75624, (Rat) 295500, (Guinea pig) 100719144, (Zebrafish) 503783
Molecular Function:
hydrolase
metalloprotease
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.