Proteintech
Manufacturer Code:602631IG150UL
Catalog # 50-173-6360
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A4A1 |
Immunogen | MEST Fusion Protein Ag11929 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Mesoderm-specific transcript homolog protein; Mesoderm-specific transcript protein; MEST PEG1; Paternally-expressed gene 1 protein
Gene Aliases: 121a; AA408879; AI256745; MEST; PEG1
UniProt ID: (Human) Q5EB52, (Mouse) Q07646
Entrez Gene ID: (Human) 4232, (Mouse) 17294
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.