Invitrogen
Manufacturer Code:PA515028
Catalog # PIPA515028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | His tag recombit protein (45-382aa) of human MERTK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-mer proto-oncogene tyrosine kinase; MER receptor tyrosine kinase; Proto-oncogene c-Mer; Proto-oncogene tyrosine-protein kinase MER Receptor tyrosine kinase MerTK MER; Receptor tyrosine kinase MerTK; STK kinase; Tyrosine-protein kinase Mer
Gene Aliases: c-Eyk; c-mer; MER; MERTK; RP38; Tyro12
UniProt ID: (Human) Q12866
Entrez Gene ID: (Human) 10461
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.