Invitrogen
Manufacturer Code:PA547460
Catalog # PIPA547460
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit human MEPE/OF45 Pro18-Asp525 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Matrix extracellular phosphoglycoprotein; matrix, extracellular phosphoglycoprotein with ASARM motif (bone); OF45; OF45 Osteoregulin extracellular phosphoglycoprotein with ASARM motif (bone) matrix extracellular phosphoglycoprotein OF45; Osteoblast/osteocyte factor 45; Osteoregulin
Gene Aliases: MEPE; OF45
UniProt ID: (Human) Q9NQ76
Entrez Gene ID: (Human) 56955
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.