Novus Biologicals
Manufacturer Code:NBP187734
Catalog # NBP187734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:KDREQRGQENTSGFWLNRSNELIWLELQAWHAGRTINDQDFFLYTARQAIPDIINEILTFKVDYGSFAFVRDRAGFNGTSVEGQCKATPGTKIVGYSTHHEHLQRQRVSFEQVKRIMELLEYIEALYPSLQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ473J16.1 (mitogen-activated protein kinase kinase kinase 4); EC 2.7.11.25 FLJ42439 MAPKKK4yeast homolog of MEKK 4 mitogen-activated protein kinase kinase kinase 4; MAP three kinase 1; MAP/ERK kinase kinase 4; MAPK/ERK kinase kinase 4; MEK kinase 4; Mitogen-activated protein kinase kinase kinase 4; predicted protein of HQ0412; SSK2/SSK22 MAP kinase kinase kinase, yeast, homolog of
Gene Aliases: KIAA0213; MAP3K4; MAPKKK4; MEKK 4; MEKK4; MTK1; PRO0412
UniProt ID: (Human) Q9Y6R4
Entrez Gene ID: (Human) 4216
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.