Invitrogen
Manufacturer Code:PA578058
Catalog # PIPA578058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human MEKK3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAP/ERK kinase kinase 3; MAPK/ERK kinase kinase 3; MAPKKK3 MEKK3 MAP3K3 MAP/ERK kinase kinase 3 MEKK 3 mitogen-activated protein kinase kinase kinase 3 MEK kinase 3 MAPK/ERK kinase kinase 3; MEK kinase 3; MEKK 3; mitogen activated protein kinase kinase kinase 3; Mitogen-activated protein kinase kinase kinase 3
Gene Aliases: AW548911; MAP3K3; MAPKKK3; MEKK3; mKIAA4031
UniProt ID: (Human) Q99759, (Mouse) Q61084
Entrez Gene ID: (Human) 4215, (Mouse) 26406
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.