Novus Biologicals
Manufacturer Code:NBP247810AF647
Catalog # NB247810647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F6 |
Immunogen | Partial recombinant MAP3K1 (aa1211-1310) (SKNSMTLDLNSSSKCDDSFGCSSNSSNAVIPSDETVFTP-VEEKCRLDVNTELNSSIEDLLEASMPSSDTTVTFKSEVAVLSPEKAENDDTYKDDVNHNQK) |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: MAP/ERK kinase kinase 1; Map3k1 MAPK/ERK kinase kinase 1 MAPKKK1MAP/ERK kinase kinase 1 MEK kinase 1 MEKK1EC 2.7.11.25 MEKKMEKK 1 mitogen-activated protein kinase kinase kinase 1; MAPK/ERK kinase kinase 1; MEK kinase 1; Mitogen-activated protein kinase kinase kinase 1; mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin protein ligase
Gene Aliases: MAP3K1; MAPKKK1; MEKK; MEKK 1; MEKK1; SRXY6
UniProt ID: (Human) Q13233
Entrez Gene ID: (Human) 4214
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.