Invitrogen
Manufacturer Code:PA543209
Catalog # PIPA543209
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human MAP3K1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAP/ERK kinase kinase 1; MAPK/ERK kinase kinase 1; MEK kinase 1; MEKK 1; mitogen activated protein kinase kinase kinase 1; Mitogen-activated protein kinase kinase kinase 1; Mitogen-activated protein kinase kinase kinase 1 E3 ubiquitin protein ligase MAPKKK1 MEKK MEKK1 SRXY6; mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin protein ligase
Gene Aliases: MAP3K1; MAPKKK1; MEKK; MEKK 1; MEKK1; si:rp71-80a19.2; SRXY6
UniProt ID: (Human) Q13233, (Rat) Q62925
Entrez Gene ID: (Human) 4214, (Dog) 478061, (Bovine) 523962, (Horse) 100061688, (Mouse) 26401, (Rat) 116667, (Zebrafish) 100334486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.