Invitrogen
Manufacturer Code:PA5111232
Catalog # PIPA5111232
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human MEKK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOS_19750 MAP3K1 MAPK/ERK kinase kinase 1 MAPKKK1 MEK kinase 1 MEKK MEKK 1 MEKK1 Mitogen-activated protein kinase kinase kinase 1 si:rp71-80a19.2 SRXY6; MAP/ERK kinase kinase 1; MAPK/ERK kinase kinase 1; MEK kinase 1; Mitogen-activated protein kinase kinase kinase 1; mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin protein ligase
Gene Aliases: MAP3K1; MAPKKK1; MEKK; MEKK 1; MEKK1; SRXY6
UniProt ID: (Human) Q13233
Entrez Gene ID: (Human) 4214
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.