Invitrogen
Manufacturer Code:MA519711
Catalog # PIMA519711
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E5 |
Immunogen | MAP2K5 (AAH08838 1 a.a. &bsim 120 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Dual specificity mitogen-activated protein kinase kinase 5; Dual specificity mitogen-activated protein kinase kinase 5 MAP kinase kinase 5 MAPKK 5 MAPK/ERK kinase 5 MAP2K5 MEK5 MKK5 PRKMK5; MAP kinase kinase 5; MAP kinase kinase MEK5b; MAPK/ERK kinase 5; MAPKK 5; MEK 5
Gene Aliases: HsT17454; MAP2K5; MAPKK5; MEK5; MKK5; PRKMK5
UniProt ID: (Human) Q13163
Entrez Gene ID: (Human) 5607
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.