Invitrogen
Manufacturer Code:PA587287
Catalog # PIPA587287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human MAP2K5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dual specificity mitogen-activated protein kinase kinase 5; Dual specificity mitogen-activated protein kinase kinase 5 MAP kinase kinase 5 MAPKK 5 MAPK/ERK kinase 5 MEK 5 MAP2K5 MEK5 MKK5 PRKMK5; MAP kinase kinase 5; MAP kinase kinase MEK5b; MAPK/ERK kinase 5; MAPK/ERK kinese 5; MAPKK 5; MEK 5; mitogen activated protein kinase kinase 5
Gene Aliases: AI324775; AI428457; HsT17454; MAP2K5; MAPKK5; MEK5; MKK5; PRKMK5
UniProt ID: (Human) Q13163, (Mouse) Q9WVS7, (Rat) Q62862
Entrez Gene ID: (Human) 5607, (Mouse) 23938, (Rat) 29568
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.