Novus Biologicals
Manufacturer Code:NBP147833
Catalog # NBP147833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F5 |
Immunogen | Full-length protein expressed in 293T cell transfected with human MAP2K1 expression vector |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Dual specificity mitogen-activated protein kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 1 ERK activator kinase 1 MAP kinase kinase 1 MAPK/ERK kinase 1 MAPKK1 MEK1MAPKK 1 mitogen-activated protein kinase kinase 1 MKK1MEK 1 PRKMK1EC 2.7.12.2 protein kinase mitogen-activated kinase 1 (MAP kinase kinase 1); ERK activator kinase 1; MAP kinase kinase 1; MAPK/ERK kinase 1; MAPKK 1; MEK 1; protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1)
Gene Aliases: CFC3; MAP2K1; MAPKK1; MEK1; MKK1; PRKMK1
UniProt ID: (Human) Q02750
Entrez Gene ID: (Human) 5604
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.