Invitrogen
Manufacturer Code:PA519311
Catalog # PIPA519311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QSNRAGFLLDPSVSQ) corresponding to the internal amino acids of MEIS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2; Homeobox protein Meis2; Meis1, myeloid ecotropic viral integration site 1 homolog 2; Meis1-related protein 1; Meis2 homeobox; MEIS2 Meis homeobox 2 HsT18361 MGC2820 MRG1 Meis homolog 2 Meis1 myeloid ecotropic viral integration site 1 homolog 2 Meis1-related gene 1 OTTHUMP00000160209 TALE homeobox protein Meis2; myeloid ecotropic viral integration site-related gene 1
Gene Aliases: A430109D20Rik; Meis2; Mrg1; Stra10
UniProt ID: (Mouse) P97367
Entrez Gene ID: (Mouse) 17536, (Rat) 311311
Molecular Function:
DNA-directed RNA polymerase
RNA binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.