Novus Biologicals
Manufacturer Code:NBP298816H
Catalog # NB055777
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human MEIS2. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein Meis2; Meis (mouse) homolog 2 Meis homeobox 2 Meis1 myeloid ecotropic viral integration site 1 homolog 2 (mouse) Meis1-related gene 1 MGC2820 myeloid ecotropic viral integration site 1 homolog 2 TALE homeobox protein Meis2; Meis homolog 2; Meis1, myeloid ecotropic viral integration site 1 homolog 2; Meis1-related gene 1; Meis1-related protein 1; TALE homeobox protein Meis2
Gene Aliases: HsT18361; MEIS2; MRG1
UniProt ID: (Human) O14770
Entrez Gene ID: (Human) 4212
Molecular Function: DNA-directed RNA polymerase RNA binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.