Proteintech
Manufacturer Code:115501AP150UL
Catalog # 50-172-7258
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MEIS2 Fusion Protein Ag2096 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2; Homeobox protein Meis2; Homeobox protein Meis2 HsT18361 Meis homeobox 2 Meis1 related protein 1 MEIS2 MRG1; Meis homolog 2; Meis1, myeloid ecotropic viral integration site 1 homolog 2; Meis1-related gene 1; Meis1-related protein 1; Meis2 homeobox; myeloid ecotropic viral integration site-related gene 1; TALE homeobox protein Meis2
Gene Aliases: A430109D20Rik; HsT18361; MEIS2; MRG1; Stra10
UniProt ID: (Human) O14770, (Mouse) P97367
Entrez Gene ID: (Human) 4212, (Mouse) 17536, (Rat) 311311
Molecular Function:
DNA-directed RNA polymerase
RNA binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.