Invitrogen
Manufacturer Code:PA570117
Catalog # PIPA570117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human C16orf73 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MEIOB C16orf73; meiosis specific with OB domains; Meiosis-specific with OB domain-containing protein; MLZ-675; Protein expressed in male leptotene and zygotene spermatocytes 675
Gene Aliases: 4930528F23Rik; C16orf73; C6H16orf73; gs129; MEIOB; MLZ-675
UniProt ID: (Human) Q8N635, (Mouse) Q9D513, (Rat) B0BMX9
Entrez Gene ID: (Horse) 100067958, (Human) 254528, (Rabbit) 100345655, (Dog) 100685856, (Mouse) 75178, (Rat) 685099
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.