Abnova
Manufacturer Code:PAB17776
Catalog # 89-119-442
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human MEGF8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of MEGF8.
For Research Use Only
Protein Aliases: C19orf49; EGF-like domain-containing protein 4; EGF-like protein 4; EGF-like-domain, multiple 4; EGFL4; Epidermal growth factor-like protein 4; FLJ22365; HBV pre-s2 binding protein 1; HBV pre-S2-binding protein 1; hepatitis B virus pre-S2-binding protein 1; MGC120684; MGC138147; Multiple EGF-like domains protein 8; multiple EGF-like-domains 8; Multiple epidermal growth factor-like domains protein 8; SBP1
Gene Aliases: C19orf49; CRPT2; EGFL4; KIAA0817; MEGF8; SBP1
UniProt ID: (Human) Q7Z7M0
Entrez Gene ID: (Human) 1954
Molecular Function: G-protein enzyme modulator extracellular matrix linker protein extracellular matrix protein receptor small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.