Invitrogen
Manufacturer Code:PA541505
Catalog # PIPA541505
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human MEF2D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MADS box transcription enhancer factor 2, polypeptide D; Myocyte enhancer factor 2D DKFZp686I1536; Myocyte-specific enhancer factor 2D
Gene Aliases: C80750; MEF2D
UniProt ID: (Human) Q14814, (Rat) O89038, (Mouse) Q63943
Entrez Gene ID: (Horse) 100064234, (Dog) 490413, (Bovine) 100336730, (Human) 4209, (Rabbit) 100340589, (Rat) 81518, (Guinea pig) 100723951, (Mouse) 17261
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.