Novus Biologicals
Manufacturer Code:NBP180566
Catalog # NBP180566
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 425 and 475 of human Myocyte Enhancer Factor 2D using the numbering given in entry NP_005911.1 (GeneID 4209). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686I1536 MADS box transcription enhancer factor 2 polypeptide D (myocyte enhancerfactor 2D) MEF2D/DAZAP1 fusion myocyte enhancer factor 2D myocyte enhancer factor 2D/deleted in azoospermia associated protein 1 fusionprotein myocyte-specific enhancer factor 2D; MADS box transcription enhancer factor 2, polypeptide D; Myocyte-specific enhancer factor 2D
Gene Aliases: MEF2D
UniProt ID: (Human) Q14814
Entrez Gene ID: (Human) 4209
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.