Invitrogen
Manufacturer Code:PA586220
Catalog # PIPA586220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 380-330 of Human MEF-2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancer factor 2A); Myocyte-specific enhancer factor 2A; Myocyte-specific enhancer factor 2A Serum response factor-like protein 1 MEF2A MEF2; Serum response factor-like protein 1
Gene Aliases: A430079H05Rik; ADCAD1; MEF2; MEF2A; RSRFC4; RSRFC9
UniProt ID: (Human) Q96D14, (Mouse) Q60929, (Rat) Q2MJT0
Entrez Gene ID: (Human) 4205, (Mouse) 17258, (Rat) 309957
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.