Invitrogen
Manufacturer Code:PA568821
Catalog # PIPA568821
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of mouse MEF2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADCAD1 RSRFC4 RSRFC9 mef2 MADS box transcription enhancer factor 2 polypeptide A (myocyte enhancer factor 2A) myocyte-specific enhancer factor 2A serum response factor-like protein 1; MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancer factor 2A); Myocyte-specific enhancer factor 2A; Serum response factor-like protein 1
Gene Aliases: A430079H05Rik; ADCAD1; MEF2; MEF2A; RSRFC4; RSRFC9
UniProt ID: (Human) Q96D14, (Mouse) Q60929, (Rat) Q2MJT0
Entrez Gene ID: (Horse) 100050068, (Goat) 102180703, (Human) 4205, (Mouse) 17258, (Rat) 309957, (Guinea pig) 100715067
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.