Invitrogen
Manufacturer Code:PA540845
Catalog # PIPA540845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human MED6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activator-recruited cofactor 33 kDa component; ARC33; CTD-2540L5.5; hMed6; Mediator complex subunit 6; Mediator complex subunit 6 NY-REN-28; mediator of RNA polymerase II transcriptio subunit 6-like protein; Mediator of RNA polymerase II transcription subunit 6; mediator of RNA polymerase II transcription, subunit 6 homolog; Renal carcinoma antigen NY-REN-28
Gene Aliases: 1500012F11Rik; ARC33; AU019660; AV213303; MED6; NY-REN-28
UniProt ID: (Human) O75586, (Bovine) Q3SZY9, (Mouse) Q921D4
Entrez Gene ID: (Human) 10001, (Dog) 490763, (Horse) 100054092, (Rabbit) 100339108, (Bovine) 505293, (Rat) 299180, (Guinea pig) 100725663, (Mouse) 69792
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.