Invitrogen
Manufacturer Code:PA520572
Catalog # PIPA520572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide from near the center of human MED28. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endothelial-derived; endothelial-derived gene 1; Endothelial-derived protein 1; FKSG20 1500003D12Rik EG1 magicin endothelial-derived gene 1 endothelial-derived protein 1 mediator of RNA polymerase II transcription subunit 28 mediator of RNA polymerase II transcription subunit 28 homolog merlin and Grb2-interacting cytoskeletal protein tumor angiogenesis marker EG-1; Magicin; Mediator complex subunit 28; Mediator of RNA polymerase II transcription subunit 28; mediator of RNA polymerase II transcription, subunit 28 homolog; Merlin and Grb2-interacting cytoskeletal protein; Tumor angiogenesis marker EG-1
Gene Aliases: 1500003D12Rik; AI451633; AU045690; EG1; FKSG20; magicin; MED28
UniProt ID: (Human) Q9H204, (Mouse) Q920D3
Entrez Gene ID: (Human) 80306, (Mouse) 66999
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.